4-Iodoimidazole ≥98.0% (by HPLC, titration analysis)

Supplier: TCI America

I0812-1G I0812-5G
TCI0812-1GEA 63.2 CAD
TCI0812-1G TCI0812-5G
4-Iodoimidazole ≥98.0% (by HPLC, titration analysis)
4-Iodoimidazole

CAS Number: 71759-89-2
MDL Number: MFCD01632213
Molecular Formula: C3H3IN2
Molecular Weight: 193.98
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal
Melting point (°C): 137

Formula: C₃H₃IN₂
Melting Pt: 137 °C
MDL Number: MFCD01632213
CAS Number: 71759-89-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=bef58231c752a03f9487af79e155893a1d82f282&vid=bef58231c752a03f9487af79e155893a1d82f282&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR