4-Iodobenzyl alcohol ≥99.0%

Supplier: TCI America

Synonyms: (4-Iodophenyl)methanol

I0810-5G I0810-25G
TCI0810-5GEA 99.22 CAD
TCI0810-5G TCI0810-25G
4-Iodobenzyl alcohol ≥99.0%
4-Iodobenzyl alcohol

CAS Number: 18282-51-4
MDL Number: MFCD01732720
Molecular Formula: C7H7IO
Molecular Weight: 234.04
Purity/Analysis Method: >99.0% (GC)
Form: Crystal
Boiling point (°C): 136
Melting point (°C): 73

Formula: C₇H₇IO
Boiling Pt: 136 °C
Melting Pt: 73 °C
MDL Number: MFCD01732720
CAS Number: 18282-51-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=25e492511591c0e1e690201ec3db1d8b29121c22&vid=25e492511591c0e1e690201ec3db1d8b29121c22&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 99.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR