2-Formylimidazole ≥98.0% (by HPLC, titration analysis)

Supplier: TCI America

I0809-5G I0809-1G
TCI0809-5GEA 158 CAD
TCI0809-5G TCI0809-1G
2-Formylimidazole ≥98.0% (by HPLC, titration analysis)
2-Formylimidazole

CAS Number: 10111-08-7
MDL Number: MFCD00003544
Molecular Formula: C4H4N2O
Molecular Weight: 96.09
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal
Melting point (°C): 202
Storage Temperature: 0-10°C


Other Product Names / Synonyms: 2-Formylimidazole

Formula: C₄H₄N₂O
Melting Pt: 202 °C
MDL Number: MFCD00003544
CAS Number: 10111-08-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9ca3d5751607f03ce7c696109c3f0310afe1303b&vid=9ca3d5751607f03ce7c696109c3f0310afe1303b&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR