Isophorone oxide ≥97.0% (by GC)
Supplier: TCI America
I0805-25G
I0805-5G
TCI0805-25GEA
305.73
CAD
TCI0805-25G
TCI0805-5G
Isophorone oxide ≥97.0% (by GC)
Isophorone oxide
CAS Number: 10276-21-8
MDL Number: MFCD00085465
Molecular Formula: C9H14O2
Molecular Weight: 154.21
Purity/Analysis Method: >97.0% (GC)
Form: Clear Liquid
Boiling point (°C): 80
Flash Point (°C): 83
Specific Gravity (20/20): 1.00
Other Product Names / Synonyms: 3,5,5-Trimethyl-2,3-epoxy-1-cyclohexanone, 2,3-Epoxy-3,5,5-trimethyl-1-cyclohexanone, 4,4,6-Trimethyl-7-oxabicyclo[4.1.0]heptan-2-one
Formula:
C₉H₁₄O₂ Boiling Pt: 80 °C Density: 1.00 (20 °C) Flash Pt: 83 °C |
MDL Number:
MFCD00085465 CAS Number: 10276-21-8 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...