2-Iodo-3-nitrotoluene ≥98.0% (by GC)

Supplier: TCI America

Synonyms: 2-Iodo-1-methyl-3-nitrobenzene

I0799-25G I0799-5G
TCI0799-25GEA 737.86 CAD
TCI0799-25G TCI0799-5G
2-Iodo-3-nitrotoluene ≥98.0% (by GC)
2-Iodo-3-nitrotoluene

CAS Number: 6277-17-4
MDL Number: MFCD00091040
Molecular Formula: C7H6INO2
Molecular Weight: 263.03
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 67


Other Product Names / Synonyms: 2-Iodo-1-methyl-3-nitrobenzene

Formula: C₇H₆INO₂
MW: 263.0300 g/mol
Melting Pt: 67 °C
MDL Number: MFCD00091040
CAS Number: 6277-17-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=23d03c74fe03101e88de08db2749e0c0ca4ecc3c&vid=23d03c74fe03101e88de08db2749e0c0ca4ecc3c&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR