2-Iodo-4-(trifluoromethyl)aniline ≥98.0%

Supplier: TCI America

Synonyms: 4-Amino-3-iodobenzotrifluoride, 2-Iodo-α,α,α-trifluoro-p-toluidine, 4-Amino-3-iodo-α,α,α-trifluorotoluene

I0794-25G I0794-5G
TCI0794-25GEA 363.4 CAD
TCI0794-25G TCI0794-5G
2-Iodo-4-(trifluoromethyl)aniline ≥98.0%
2-Iodo-4-(trifluoromethyl)aniline

CAS Number: 163444-17-5
MDL Number: MFCD02683097
Molecular Formula: C7H5F3IN
Molecular Weight: 287.02
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 52


Other Product Names / Synonyms: 4-Amino-3-iodobenzotrifluoride

Formula: C₇H₅F₃IN
MW: 287.02
Melting Pt: 52 °C
MDL Number: MFCD02683097
CAS Number: 163444-17-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=dd17f1ffef0be689f1dd6d1893b3b235e6a64736&vid=dd17f1ffef0be689f1dd6d1893b3b235e6a64736&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR