4-Iodo-1-methylpyrazole-5-carboxylic acid ≥98.0%

Supplier: TCI America

Synonyms: 4-Iodo-1-methyl-1H-pyrazole-5-carboxylic acid

I0786-1G I0786-5G
TCI0786-1GEA 105.86 CAD
TCI0786-1G TCI0786-5G
4-Iodo-1-methylpyrazole-5-carboxylic acid ≥98.0%
4-Iodo-1-methylpyrazole-5-carboxylic acid

CAS Number: 75092-30-7
MDL Number: MFCD00461121
Molecular Formula: C5H5IN2O2
Molecular Weight: 252.01
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Melting point (°C): 242

Formula: C₅H₅IN₂O₂
MW: 252.01 g/mol
Melting Pt: 242 °C
MDL Number: MFCD00461121
CAS Number: 75092-30-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a7e09bdd19c20ada8b665bc1097df079e24cce37&vid=a7e09bdd19c20ada8b665bc1097df079e24cce37&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR