Iron(II) phthalocyanine ≥98.0% (by titrimetric analysis), purified by sublimation

Supplier: TCI America

Synonyms: Phthalocyanine Iron(II), Iron phthalocyanine

I0783-1G I0783-200MG
TCI0783-1GEA 542.1 CAD
TCI0783-1G TCI0783-200MG
Iron(II) phthalocyanine ≥98.0% (by titrimetric analysis), purified by sublimation
Iron(II) phthalocyanine

CAS Number: 132-16-1
MDL Number: MFCD00015953
Molecular Formula: C32H16FeN8
Molecular Weight: 568.38
Purity/Analysis Method: >98.0% (T)
Form: Crystal


Product Categories: Materials Chemistry, Molecular Conductors, Porphyrin & Phthalocyanine Dyes (Functional Organic Dyes), Highly Purified Reagents, Building Blocks for Phthalocyanines
Other Product Names / Synonyms: Phthalocyanine Iron(II)

Formula: C₃₂H₁₆FeN₈
MW: 568.38 g/mol
MDL Number: MFCD00015953
CAS Number: 132-16-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=459012f0e78e6d48ba44545f9065d085803797f4&vid=7977bb38686bd7f09fa1acca9bc19220cb60b9cf&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR