2-(4-Iodophenyl)-3-(4-nitrophenyl)-5-phenyltetrazolium chloride ≥98.0% (by HPLC) for biochemical research

Supplier: TCI America

I0781-100MG I0781-1G
TCI0781-100MGEA 28.44 CAD
TCI0781-100MG TCI0781-1G
2-(4-Iodophenyl)-3-(4-nitrophenyl)-5-phenyltetrazolium chloride ≥98.0% (by HPLC) for biochemical research
2-(4-Iodophenyl)-3-(4-nitrophenyl)-5-phenyltetrazolium chloride

[for Biochemical Research]
CAS Number: 146-68-9
MDL Number: MFCD00011961
Molecular Formula: C19H13ClIN5O2
Molecular Weight: 505.70
Purity/Analysis Method: >98.0% (HPLC)
Form: Crystal
Storage Temperature: 0-10°C


Product Categories: Bioscience, Alkali Phosphatase Substrates
Other Product Names / Synonyms: INT, Iodonitrotetrazolium Chloride, p-Iodonitrotetrazolium Violet

Formula: C₁₉H₁₃ClIN₅O₂
MDL Number: MFCD00011961
CAS Number: 146-68-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f2fdf454876bb756601be375474d02002e813fb3&vid=ef9ccb6bfc48f57117961babb80aea5f668d4d31&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR