3,5-Dimethyl-4-iodo-1H-pyrazole ≥98.0%

Supplier: TCI America

Synonyms: 3,5-Dimethyl-4-iodopyrazole

I0771-1G I0771-5G
TCI0771-1GEA 33.18 CAD
TCI0771-1G TCI0771-5G
3,5-Dimethyl-4-iodo-1H-pyrazole ≥98.0%
3,5-Dimethyl-4-iodo-1H-pyrazole

CAS Number: 2033-45-6
MDL Number: MFCD00040247
Molecular Formula: C5H7IN2
Molecular Weight: 222.03
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 140
Lambda max.: 223 nm (EtOH)

Formula: C₅H₇IN₂
MW: 222.03
Melting Pt: 140 °C
MDL Number: MFCD00040247
CAS Number: 2033-45-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8866983f01d3a326a4532180db12a8c5bbcb5c7a&vid=8866983f01d3a326a4532180db12a8c5bbcb5c7a&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR