Imidazolidinyl urea

Supplier: TCI America

I0665-250G I0665-25G
TCI0665-250GEA 271.76 CAD
TCI0665-250G TCI0665-25G
Imidazolidinyl urea
Imidazolidinyl urea

CAS Number: 39236-46-9
MDL Number: MFCD00221482
Molecular Formula: C11H16N8O8
Molecular Weight: 388.30
Form: Crystal
Color: White
Storage Temperature: 0-10°C


Other Product Names / Synonyms: N,N'-Methylenebis[N'-(3-hydroxymethyl-2,5-dioxo-4-imidazolidinyl)urea], Imidurea

Formula: C₁₁H₁₆N₈O₈
MDL Number: MFCD00221482
CAS Number: 39236-46-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=880a72d86fbdfd9fa09e81fc84694c61ba5c9e78&vid=880a72d86fbdfd9fa09e81fc84694c61ba5c9e78&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR