Methyl-4-iodobenzoate ≥98.0% (by GC)
Supplier: TCI America
I0593-25G
I0593-5G
TCI0593-25GEA
116.92
CAD
TCI0593-25G
TCI0593-5G
Methyl-4-iodobenzoate ≥98.0% (by GC)
Methyl 4-iodobenzoate
CAS Number: 619-44-3
MDL Number: MFCD00016353
Molecular Formula: C8H7IO2
Molecular Weight: 262.05
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 114
Other Product Names / Synonyms: 4-Iodobenzoic Acid Methyl Ester
Formula:
C₈H₇IO₂ Melting Pt: 114 °C |
MDL Number:
MFCD00016353 CAS Number: 619-44-3 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...