Iodomesitylene diacetate ≥98.0% (by titrimetric analysis)

Supplier: TCI America

Synonyms: 2,4,6-Trimethyl(diacetoxyiodo)benzene, 2,4,6-Trimethyliodobenzene Diacetate, 2-(Diacetoxyiodo)mesitylene

I0479-25G I0479-5G
TCI0479-25GEA 472.01 CAD
TCI0479-25G TCI0479-5G
Iodomesitylene diacetate ≥98.0% (by titrimetric analysis)
Iodomesitylene diacetate

CAS Number: 33035-41-5
MDL Number: MFCD00674196
Molecular Formula: C13H17IO4
Molecular Weight: 364.18
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Melting point (°C): 168
Storage Temperature: 0-10°C


Product Categories: Synthetic Organic Chemistry, Hypervalent Iodine Compounds, Oxidation
Other Product Names / Synonyms: 2-(Diacetoxyiodo)mesitylene, 2,4,6-Trimethyl(diacetoxyiodo)benzene, 2,4,6-Trimethyliodobenzene Diacetate

Formula: C₁₃H₁₇IO₄
MW: 364.18 g/mol
Melting Pt: 168 °C
MDL Number: MFCD00674196
CAS Number: 33035-41-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=48d24e2f0545eed57c92a8791932ebf438d5ad4a&vid=48d24e2f0545eed57c92a8791932ebf438d5ad4a&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR