2-Iodophenol ≥99.0% (by GC)

Supplier: TCI America

I0460-25G I0460-5G
TCI0460-25GEA 213.3 CAD
TCI0460-25G TCI0460-5G
2-Iodophenol ≥99.0% (by GC)
2-Iodophenol

CAS Number: 533-58-4
MDL Number: MFCD00013963
Molecular Formula: C6H5IO
Molecular Weight: 220.01
Purity/Analysis Method: >99.0% (GC)
Form: Crystal
Boiling point (°C): 187
Melting point (°C): 42
Flash Point (°C): 230
Storage Temperature: 0-10°C

Formula: C₆H₅IO
MW: 220.01 g/mol
Boiling Pt: 187 °C
Melting Pt: 42 °C
Flash Pt: 230 °C
MDL Number: MFCD00013963
CAS Number: 533-58-4
UN: 2811
ADR: 6.1,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=817ea8833c0e8fbccc53403bfa627b3740adf7c6&vid=6e75eb771177018a50b66c05693038cced40988b&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 99.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR