4'-Isobutylacetophenone ≥96.0% (by GC)

Supplier: TCI America

Synonyms: p-Isobutylacetophenone

I0417-25ML I0417-500ML I0417-100ML
TCI0417-25MLEA 147.32 CAD
TCI0417-25ML TCI0417-500ML TCI0417-100ML
4'-Isobutylacetophenone ≥96.0% (by GC)
4'-Isobutylacetophenone

CAS Number: 38861-78-8
MDL Number: MFCD00027393
Molecular Formula: C12H16O
Molecular Weight: 176.26
Purity/Analysis Method: >96.0% (GC)
Form: Clear Liquid
Boiling point (°C): 268
Flash Point (°C): 100
Specific Gravity (20/20): 0.96

Formula: C₁₂H₁₆O
MW: 176.26 g/mol
Boiling Pt: 268 °C
Density: 0.96 (20 °C)
Flash Pt: 100 °C
MDL Number: MFCD00027393
CAS Number: 38861-78-8
UN: 1224
ADR: 3,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=cc99743cc02d304e102d41572c69c040b94e8b23&vid=cc99743cc02d304e102d41572c69c040b94e8b23&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 96.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR