Iodosobenzene diacetate ≥97.0% (by titrimetric analysis)

Supplier: TCI America

I0330-10G I0330-250G I0330-25G
TCI0330-10GEA 43.47 CAD
TCI0330-10G TCI0330-250G TCI0330-25G
Iodosobenzene diacetate ≥97.0% (by titrimetric analysis)
Iodosobenzene diacetate

CAS Number: 3240-34-4
MDL Number: MFCD00008692
Molecular Formula: C10H11IO4
Molecular Weight: 322.10
Purity/Analysis Method: >97.0% (T)
Form: Crystal
Melting point (°C): 158


Product Categories: Synthetic Organic Chemistry, Hypervalent Iodine Compounds, Oxidation
Other Product Names / Synonyms: (Diacetoxyiodo)benzene, PIDA

Formula: C₁₀H₁₁IO₄
Melting Pt: 158 °C
MDL Number: MFCD00008692
CAS Number: 3240-34-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=84a9713ccfd820832ae61477755462e0b2a5e928&vid=84a9713ccfd820832ae61477755462e0b2a5e928&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR