2-(4-Iodophenyl)-3-(4-nitrophenyl)-5-phenyltetrazolium chloride ≥98.0%

Supplier: TCI America

Synonyms: Iodonitrotetrazolium chloride, p-Iodonitrotetrazolium violet, INT, 3-(4-Iodophenyl)-2-(4-nitrophenyl)-5-phenyl-2H-tetrazolium chloride, 2-(4-Iodophenyl)-3-(4-nitrophenyl)-5-phenyl-2H-tetrazolium chloride, 2-(4-Iodophenyl)-3-(4-nitrophenyl)-5-phenyl-2H-tetrazol-3-ium chloride, Iodonitrotetrazolium violet, 3-(4-Iodophenyl)-2-(4-nitrophenyl)-5-phenyltetrazolium chloride

I0067-1G I0067-5G
TCI0067-1GEA 96.46 CAD
TCI0067-1G TCI0067-5G
2-(4-Iodophenyl)-3-(4-nitrophenyl)-5-phenyltetrazolium chloride ≥98.0%
2-(4-Iodophenyl)-3-(4-nitrophenyl)-5-phenyltetrazolium chloride

CAS Number: 146-68-9
MDL Number: MFCD00011961
Molecular Formula: C19H13ClIN5O2
Molecular Weight: 505.70
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal
Storage Temperature: 0-10°C


Product Categories: Bioscience, Stains & Dyes, Redox Research Reagents
Other Product Names / Synonyms: INT, Iodonitrotetrazolium Chloride, p-Iodonitrotetrazolium Violet

Formula: C₁₉H₁₃ClIN₅O₂
MDL Number: MFCD00011961
CAS Number: 146-68-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f24ba380efad899f625fcc97136a4dbfa23efe14&vid=f24ba380efad899f625fcc97136a4dbfa23efe14&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR