3-(Hydroxymethyl)biphenyl ≥96.0%
Supplier: TCI America
H1481-1G
H1481-5G
TCH1481-1GEA
222.78
CAD
TCH1481-1G
TCH1481-5G
3-(Hydroxymethyl)biphenyl ≥96.0%
3-(Hydroxymethyl)biphenyl
CAS Number: 69605-90-9
MDL Number: MFCD08703631
Molecular Formula: C13H12O
Molecular Weight: 184.24
Purity/Analysis Method: >96.0% (GC)
Form: Crystal
Melting point (°C): 50
Other Product Names / Synonyms: Biphenyl-3-methanol, 3-Phenylbenzyl Alcohol
Formula:
C₁₃H₁₂O Melting Pt: 50 °C |
MDL Number:
MFCD08703631 CAS Number: 69605-90-9 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...