5-Hydroxynorbornane-2,6-lactone ≥98.0% (by GC)
Supplier: TCI America
H1475-25G
H1475-5G
TCH1475-25GEA
128.93
CAD
TCH1475-25G
TCH1475-5G
5-Hydroxynorbornane-2,6-lactone ≥98.0% (by GC)
5-Hydroxynorbornane-2,6-lactone
CAS Number: 92343-46-9
MDL Number: MFCD00988184
Molecular Formula: C8H10O3
Molecular Weight: 154.17
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 158
Product Categories: Materials Chemistry, Monomers
Other Product Names / Synonyms: 6-Hydroxyhexahydro-2H-3,5-methanocyclopenta[b]furan-2-one
Formula:
C₈H₁₀O₃ Melting Pt: 158 °C |
MDL Number:
MFCD00988184 CAS Number: 92343-46-9 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...