all-trans-N-(4-Hydroxyphenyl)retinamide ≥98.0%

Supplier: TCI America

Synonyms: N-(4-Hydroxyphenyl)-all- trans retinamide, Retinoic Acid p-Hydroxyanilide, (2E,4E,6E,8E)-N-(4-Hydroxyphenyl)-3,7-dimethyl-9-(2,6,6-trimethylcyclohex-1-en-1-yl)nona-2,4,6,8-tetraenamide, all-trans-N-(4-Hydroxyphenyl)retinamide, Fenretinide, 4-HPR

H1464-100MG H1464-10MG
TCH1464-100MGEA 1096.52 CAD
TCH1464-100MG TCH1464-10MG
all-trans-N-(4-Hydroxyphenyl)retinamide ≥98.0%
4-Hydroxyphenylretinamide

CAS Number: 65646-68-6
MDL Number: MFCD00792674
Molecular Formula: C26H33NO2
Molecular Weight: 391.56
Purity/Analysis Method: >98.0% (HPLC)
Form: Crystal
Melting point (°C): 172
Lambda max.: 362 nm (MeOH)Storage Temperature: >-20°C


Product Categories: Bioscience, Antitumor Ingredients, Vitamin Derivatives
Other Product Names / Synonyms: 4-HPR, Fenretinide, Retinoic Acid p-Hydroxyanilide

Formula: C₂₆H₃₃NO₂
MW: 391.56 g/mol
Melting Pt: 172 °C
MDL Number: MFCD00792674
CAS Number: 65646-68-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a37b75f5d72bd48dc6c5469239813a4bae83726e&vid=a37b75f5d72bd48dc6c5469239813a4bae83726e&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR