3-Hexadecylthiophene ≥97.0%

Supplier: TCI America

H1462-1G H1462-25G H1462-5G
TCH1462-1GEA 48.98 CAD
TCH1462-1G TCH1462-25G TCH1462-5G
3-Hexadecylthiophene ≥97.0%
3-Hexadecylthiophene

CAS Number: 119269-24-8
MDL Number: MFCD07368973
Molecular Formula: C20H36S
Molecular Weight: 308.57
Purity/Analysis Method: >97.0% (GC)
Form: Crystal
Melting point (°C): 33
Flash Point (°C): 110
Lambda max.: 244 nm (CHCl3)

Formula: C₂₀H₃₆S
Melting Pt: 33 °C
Flash Pt: 110 °C
MDL Number: MFCD07368973
CAS Number: 119269-24-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e6a6d869c4382808b9dc132087249eaeba233d9c&vid=e6a6d869c4382808b9dc132087249eaeba233d9c&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR