(3-Hydroxy-4-methoxyphenyl)acetic acid ≥98.0% (by GC, titration analysis)

Supplier: TCI America

H1455-1G H1455-5G
TCH1455-1GEA 105.86 CAD
TCH1455-1G TCH1455-5G
(3-Hydroxy-4-methoxyphenyl)acetic acid ≥98.0% (by GC, titration analysis)
(3-Hydroxy-4-methoxyphenyl)acetic acid

CAS Number: 1131-94-8
MDL Number: MFCD00016829
Molecular Formula: C9H10O4
Molecular Weight: 182.18
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Melting point (°C): 131


Other Product Names / Synonyms: Homoisovanillic Acid

Formula: (C₆H₃(OH)(OCH₃))CH₂COOH
Melting Pt: 131 °C
MDL Number: MFCD00016829
CAS Number: 1131-94-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c8a81cfcf563561871bf5f953bf4729600ef3869&vid=c8a81cfcf563561871bf5f953bf4729600ef3869&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR