1,2,3,4,5,6-Hexa-O-[11-[4-(4-hexylphenylazo)phenoxy]undecanoyl]-D-mannitol ≥97.0% (by HPLC)
Supplier: TCI America
H1452-1G
H1452-5G
TCH1452-1GEA
593.45
CAD
TCH1452-1G
TCH1452-5G
1,2,3,4,5,6-Hexa-O-[11-[4-(4-hexylphenylazo)phenoxy]undecanoyl]-D-mannitol ≥97.0% (by HPLC)
1,2,3,4,5,6-Hexa-O-[11-[4-(4-hexylphenylazo)phenoxy]undecanoyl]-D-mannitol
CAS Number: 1093077-77-0
Molecular Formula: C180H254N12O18
Molecular Weight: 2874.08
Purity/Analysis Method: >97.0% (HPLC)
Form: Crystal
Formula:
C₁₈₀H₂₅₄N₁₂O₁₈ MW: 2874.08 g/mol |
CAS Number:
1093077-77-0 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...