2,3,6,7,10,11-Hexakis[(n-octyl)oxy]triphenylene ≥98.0% (by HPLC)

Supplier: TCI America

H1450-1G H1450-200MG
TCH1450-1GEA 1041.22 CAD
TCH1450-1G TCH1450-200MG
2,3,6,7,10,11-Hexakis[(n-octyl)oxy]triphenylene ≥98.0% (by HPLC)
2,3,6,7,10,11-Hexakis[(n-octyl)oxy]triphenylene

CAS Number: 70351-87-0
MDL Number: MFCD20528916
Molecular Formula: C66H108O6
Molecular Weight: 997.58
Purity/Analysis Method: >98.0% (HPLC)
Form: Crystal
Lambda max.: 280 nm (petroleum ether)

Formula: C₆₆H₁₀₈O₆
MW: 997.58 g/mol
MDL Number: MFCD20528916
CAS Number: 70351-87-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=43877122bcba231b550dc0588c5617850cdedede&vid=43877122bcba231b550dc0588c5617850cdedede&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR