2,3,6,7,10,11-Hexakis(hexyloxy)triphenylene ≥98.0% (by HPLC)
Supplier: TCI America
H1449-1G
H1449-200MG
TCH1449-1GEA
872.16
CAD
TCH1449-1G
TCH1449-200MG
2,3,6,7,10,11-Hexakis(hexyloxy)triphenylene ≥98.0% (by HPLC)
2,3,6,7,10,11-Hexakis(hexyloxy)triphenylene
CAS Number: 70351-86-9
MDL Number: MFCD16250223
Molecular Formula: C54H84O6
Molecular Weight: 829.26
Purity/Analysis Method: >98.0% (HPLC)
Form: Crystal
Lambda max.: 278 nm (CH2Cl2)
Formula:
C₅₄H₈₄O₆ |
MDL Number:
MFCD16250223 CAS Number: 70351-86-9 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...