2-Hydroxydimetridazole ≥98.0% (by GC, titration analysis)

Supplier: TCI America

H1439-1G H1439-5G
TCH1439-1GEA 165.9 CAD
TCH1439-1G TCH1439-5G
2-Hydroxydimetridazole ≥98.0% (by GC, titration analysis)
2-Hydroxydimetridazole

CAS Number: 936-05-0
MDL Number: MFCD00159675
Molecular Formula: C5H7N3O3
Molecular Weight: 157.13
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Melting point (°C): 117


Other Product Names / Synonyms: 1-Methyl-5-nitroimidazole-2-methanol

Formula: C₅H₇N₃O₃
Melting Pt: 117 °C
MDL Number: MFCD00159675
CAS Number: 936-05-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=cb8af96c8e16f85a2fd4aba8cd190dc7ec62b314&vid=cb8af96c8e16f85a2fd4aba8cd190dc7ec62b314&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR