3-Hydroxy-ɑ-methylbenzyl alcohol ≥98.0% (by GC)

Supplier: TCI America

H1433-1G H1433-5G
TCH1433-1GEA 148.52 CAD
TCH1433-1G TCH1433-5G
3-Hydroxy-ɑ-methylbenzyl alcohol ≥98.0% (by GC)
3-Hydroxy-alpha-methylbenzyl alcohol

CAS Number: 2415-09-0
MDL Number: MFCD00238621
Molecular Formula: C8H10O2
Molecular Weight: 138.17
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 118


Other Product Names / Synonyms: 1-(3-Hydroxyphenyl)ethanol, 3-(1-Hydroxyethyl)phenol

Formula: C₈H₁₀O₂
Melting Pt: 118 °C
MDL Number: MFCD00238621
CAS Number: 2415-09-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=6d03eaae7413424427d49f1179e133ee3707bcff&vid=6d03eaae7413424427d49f1179e133ee3707bcff&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR