Hexaethylene glycol ≥98.0%

Supplier: TCI America

Synonyms: 3,6,9,12,15-Pentaoxaheptadecane-1,17-diol

H1432-25G H1432-5G
TCH1432-25GEA 687.3 CAD
TCH1432-25G TCH1432-5G
Hexaethylene glycol ≥98.0%
Hexaethylene glycol

CAS Number: 2615-15-8
MDL Number: MFCD00002877
Molecular Formula: C12H26O7
Molecular Weight: 282.33
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 217
Melting point (°C): 7
Flash Point (°C): 113
Specific Gravity (20/20): 1.13

Formula: C₁₂H₂₆O₇
MW: 282.33 g/mol
Boiling Pt: 217 °C
Melting Pt: 7 °C
Density: 1.13 (20 °C)
Flash Pt: 113 °C
MDL Number: MFCD00002877
CAS Number: 2615-15-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1737887cd2476c11baec3ffc6fe7042ade565d0d&vid=1737887cd2476c11baec3ffc6fe7042ade565d0d&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR