1-Hexyl-3-methylimidazolium bis(trifluoromethylsulfonyl)imide ≥98.0%
Supplier: TCI America
Synonyms:
3-Hexyl-1-methyl-1H-imidazol-3-ium bis((trifluoromethyl)sulfonyl)amide, 3-Hexyl-1-methyl-1H-imidazol-3-ium bis((trifluoromethyl)sulphonyl)amide, 1-Hexyl-3-methylimidazolium Bis(trifluoromethanesulfonyl)imide, 1-Hexyl-3-methylimidazolium Bis(trifluoromethanesulphonyl)imide
CAS Number: 382150-50-7
MDL Number: MFCD08458931
Molecular Formula: C12H19F6N3O4S2
Molecular Weight: 447.41
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Clear Liquid
Melting point (°C): -9
Specific Gravity (20/20): 1.38
Formula:
C₁₂H₁₉F₆N₃O₄S₂ MW: 447.42 g/mol Melting Pt: –9 °C Density: 1.38 (20 °C) |
MDL Number:
MFCD08458931 CAS Number: 382150-50-7 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...