4-Hydroxy-6-methylcoumarin ≥98.0%
Supplier: TCI America
H1421-1G
H1421-5G
TCH1421-1GEA
48.98
CAD
TCH1421-1G
TCH1421-5G
4-Hydroxy-6-methylcoumarin ≥98.0%
4-Hydroxy-6-methylcoumarin
CAS Number: 13252-83-0
MDL Number: MFCD00085738
Molecular Formula: C10H8O3
Molecular Weight: 176.17
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 261
Lambda max.: 309 nm (EtOH aq.)
Formula:
C₁₀H₈O₃ MW: 176.17 g/mol Melting Pt: 261 °C |
MDL Number:
MFCD00085738 CAS Number: 13252-83-0 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...