(-)-Corey lactone ≥98.0% (by GC)

Supplier: TCI America

Synonyms: (-)-6β-Hydroxymethyl-7α-hydroxy-cis-2-oxabicyclo[3.3.0]octan-3-one, (3aR,4S,5R,6aS)-5-Hydroxy-4-(hydroxymethyl)hexahydro-2H-cyclopenta[b]furan-2-one, (3aR,4S,5R,6aS)-(-)-Hexahydro-5-hydroxy-4-(hydroxymethyl)-2H-cyclopenta[b]furan-2-one, (-)-Corey lactone diol

H1418-1G H1418-200MG
TCH1418-1GEA 294.2 CAD
TCH1418-1G TCH1418-200MG
(-)-Corey lactone ≥98.0% (by GC)
(-)-Corey lactone

CAS Number: 32233-40-2
MDL Number: MFCD00075234
Molecular Formula: C8H12O4
Molecular Weight: 172.18
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 116
Specific rotation [a]20/D: -41 deg (C=1, MeOH)


Product Categories: Synthetic Organic Chemistry, Chiral Building Blocks
Other Product Names / Synonyms: (3aR,4S,5R,6aS)-(-)-Hexahydro-5-hydroxy-4-(hydroxymethyl)-2H-cyclopenta[b]furan-2-one

Formula: C₈H₁₂O₄
MW: 172.18 g/mol
Melting Pt: 116 °C
MDL Number: MFCD00075234
CAS Number: 32233-40-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=6bb44fa4a1c46d82d5c8310f3e1c82b3b0a9d5b3&vid=6bb44fa4a1c46d82d5c8310f3e1c82b3b0a9d5b3&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR