3-(Hydroxymethyl)-1-adamantanol ≥98.0%

Supplier: TCI America

Synonyms: 3-(Hydroxymethyl)adamantan-1-ol, 1-Hydroxy-3-(hydroxymethyl)adamantane

H1415-1G H1415-5G
TCH1415-1GEA 55.3 CAD
TCH1415-1G TCH1415-5G
3-(Hydroxymethyl)-1-adamantanol ≥98.0%
3-(Hydroxymethyl)-1-adamantanol

CAS Number: 38584-37-1
MDL Number: MFCD03105096
Molecular Formula: C11H18O2
Molecular Weight: 182.26
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 158


Product Categories: Materials Chemistry, Monomers
Other Product Names / Synonyms: 1-Hydroxy-3-(hydroxymethyl)adamantane

Formula: C₁₁H₁₈O₂
Melting Pt: 158 °C
MDL Number: MFCD03105096
CAS Number: 38584-37-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a3f580c98963577fc6f7020d9c60bab6f61f0fc2&vid=a3f580c98963577fc6f7020d9c60bab6f61f0fc2&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR