3-Hydroxy-1-adamantaneacetic acid ≥97.0%

Supplier: TCI America

Synonyms: 1-(Carboxymethyl)-3-hydroxyadamantane, 2-(3-Hydroxyadamantan-1-yl)acetic acid

H1414-1G H1414-5G
TCH1414-1GEA 105.86 CAD
TCH1414-1G TCH1414-5G
3-Hydroxy-1-adamantaneacetic acid ≥97.0%
3-Hydroxy-1-adamantaneacetic acid

CAS Number: 17768-36-4
MDL Number: MFCD00167797
Molecular Formula: C12H18O3
Molecular Weight: 210.27
Purity/Analysis Method: >97.0% (T)
Form: Crystal
Melting point (°C): 125


Other Product Names / Synonyms: 1-(Carboxymethyl)-3-hydroxyadamantane

Formula: C₁₂H₁₈O₃
Melting Pt: 125 °C
MDL Number: MFCD00167797
CAS Number: 17768-36-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e60c1929847e617dec29cc6527969ca346e14405&vid=e60c1929847e617dec29cc6527969ca346e14405&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR