2-Hydroxycyclohexanecarboxylic acid ≥98.0% (by GC, titration analysis)

Supplier: TCI America

H1409-1G H1409-5G
TCH1409-1GEA 369.72 CAD
TCH1409-1G TCH1409-5G
2-Hydroxycyclohexanecarboxylic acid ≥98.0% (by GC, titration analysis)
2-Hydroxycyclohexanecarboxylic acid

CAS Number: 609-69-8
MDL Number: MFCD02684437
Molecular Formula: C7H12O3
Molecular Weight: 144.17
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Melting point (°C): 69

Formula: C₇H₁₂O₃
Melting Pt: 69 °C
MDL Number: MFCD02684437
CAS Number: 609-69-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8c1d983e07bc2b2347c3c515f4108a2ab35a5204&vid=8c1d983e07bc2b2347c3c515f4108a2ab35a5204&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR