3-Hydroxycyclohexanecarboxylic acid (cis and trans mixture) ≥98.0% (by GC, titration analysis)

Supplier: TCI America

Synonyms: 3-Hydroxycyclohexanecarboxylic acid, 3-Hydroxycyclohexane-1-carboxylic acid

H1408-1G H1408-5G
TCH1408-1GEA 437.66 CAD
TCH1408-1G TCH1408-5G
3-Hydroxycyclohexanecarboxylic acid (cis and trans mixture) ≥98.0% (by GC, titration analysis)
3-Hydroxycyclohexanecarboxylic acid (cis and trans mixture)

CAS Number: 606488-94-2
Molecular Formula: C7H12O3
Molecular Weight: 144.17
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal

Formula: C₇H₁₂O₃
MW: 144.17 g/mol
MDL Number: MFCD19228828
CAS Number: 606488-94-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4a1dfffaa4febddbf943f47bbb3497bd403b74c1&vid=4a1dfffaa4febddbf943f47bbb3497bd403b74c1&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR