(2S)-N-[(1S)-1-(Hydroxydiphenylmethyl)-3-methylbutyl]-2-pyrrolidinecarboxamide ≥98.0%

Supplier: TCI America

Synonyms: N-[(1S)-1-(Hydroxydiphenylmethyl)-3-methylbutyl]-L-prolinamide, Singh's Catalyst, (S)-N-[(S)-1-Hydroxy-4-methyl-1,1-diphenylpentan-2-yl]pyrrolidine-2-carboxamide

H1407-1G H1407-200MG
TCH1407-1GEA 697.57 CAD
TCH1407-1G TCH1407-200MG
(2S)-N-[(1S)-1-(Hydroxydiphenylmethyl)-3-methylbutyl]-2-pyrrolidinecarboxamide ≥98.0%
(2S)-N-[(1S)-1-(Hydroxydiphenylmethyl)-3-methylbutyl]-2-pyrrolidinecarboxamide

CAS Number: 910110-45-1
MDL Number: MFCD22200506
Molecular Formula: C23H30N2O2
Molecular Weight: 366.51
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal
Melting point (°C): 193
Specific rotation [a]20/D: -47 deg (C=1, CHCl3)


Product Categories: Synthetic Organic Chemistry, Asymmetric Organocatalysts
Other Product Names / Synonyms: (S)-N-[(S)-1-Hydroxy-4-methyl-1,1-diphenylpentan-2-yl]pyrrolidine-2-carboxamide, N-[(1S)-1-(Hydroxydiphenylmethyl)-3-methylbutyl]-L-prolinamide, Singh's Catalyst

Formula: C₂₃H₃₀N₂O₂
Melting Pt: 193 °C
MDL Number: MFCD22200506
CAS Number: 910110-45-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=88d0403dfa604574c57a839e55285e556470d8d8&vid=88d0403dfa604574c57a839e55285e556470d8d8&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR