3-Hydroxy-4'-methoxyflavone ≥98.0%

Supplier: TCI America

Synonyms: 3-Hydroxy-2-(4-methoxyphenyl)-4H-chromen-4-one, 4'-Methoxyflavonol, 3-Hydroxy-2-(4-methoxyphenyl)-4-benzopyrone, 3-Hydroxy-2-(4-methoxyphenyl)chromen-4-one

H1405-1G H1405-5G
TCH1405-1GEA 134.3 CAD
TCH1405-1G TCH1405-5G
3-Hydroxy-4'-methoxyflavone ≥98.0%
3-Hydroxy-4'-methoxyflavone

CAS Number: 6889-78-7
MDL Number: MFCD00017682
Molecular Formula: C16H12O4
Molecular Weight: 268.27
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 234
Lambda max.: 350 nm (EtOH)


Product Categories: Bioscience, Aromatic Polyketides
Other Product Names / Synonyms: 3-Hydroxy-2-(4-methoxyphenyl)chromone

Formula: C₁₆H₁₂O₄
Melting Pt: 234 °C
MDL Number: MFCD00017682
CAS Number: 6889-78-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ffdd30752c1204701a4016e0e34d993fe44d0abb&vid=ffdd30752c1204701a4016e0e34d993fe44d0abb&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR