2-Hydroxy-2-azaadamantane ≥98.0% (by GC, titration analysis)

Supplier: TCI America

H1404-1G H1404-200MG H1404-5G
TCH1404-1GEA 490.91 CAD
TCH1404-1G TCH1404-200MG TCH1404-5G
2-Hydroxy-2-azaadamantane ≥98.0% (by GC, titration analysis)
2-Hydroxy-2-azaadamantane

CAS Number: 1155843-79-0
Molecular Formula: C9H15NO
Molecular Weight: 153.23
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal


Product Categories: Synthetic Organic Chemistry, Oxidation, Oxidation
Other Product Names / Synonyms: AZADOL(regR)

Formula: C₉H₁₅NO
CAS Number: 1155843-79-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=0a62d99a088da4c0ffa711a32a83b7f494ed4c78&vid=0a62d99a088da4c0ffa711a32a83b7f494ed4c78&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR