6-Hydroxy-2-naphthaleneboronic acid (contains varying amounts of Anhydride)
Supplier: TCI America
H1402-1G
H1402-5G
TCH1402-1GEA
91.64
CAD
TCH1402-1G
TCH1402-5G
6-Hydroxy-2-naphthaleneboronic acid (contains varying amounts of Anhydride)
6-Hydroxy-2-naphthaleneboronic acid
CAS Number: 173194-95-1
MDL Number: MFCD00092986
Molecular Formula: C10H9BO3
Molecular Weight: 187.99
Form: Crystal
Color: Slightly Pale Yellow
Melting point (°C): 224
Other Product Names / Synonyms: 6-Hydroxy-2-naphthylboronic Acid
Formula:
C₁₀H₉BO₃ Melting Pt: 224 °C |
MDL Number:
MFCD00092986 CAS Number: 173194-95-1 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...