(2-Hydroxyethyl)dimethyl(3-sulfopropyl)ammonium hydroxide inner salt 98,0 HPLC, N Biochemical Research

Supplier: TCI America

Synonyms: 3-((2-Hydroxyethyl)dimethylammonio)propane-1-sulfonate, 3-[(2-Hydroxyethyl)dimethylammonio]propane-1-sulphonate, 3-((2-Hydroxyethyl)dimethylammonio)propane-1-sulphonate, 3-[(2-Hydroxyethyl)dimethylammonio]propane-1-sulfonate, NDSB 211

H1399-1G H1399-5G
TCH1399-1GEA 128.93 CAD
TCH1399-1G TCH1399-5G
(2-Hydroxyethyl)dimethyl(3-sulfopropyl)ammonium hydroxide inner salt 98,0 HPLC, N Biochemical Research
(2-Hydroxyethyl)dimethyl(3-sulfopropyl)ammonium hydroxide inner salt

[for Biochemical Research]
CAS Number: 38880-58-9
MDL Number: MFCD01320397
Molecular Formula: C7H17NO4S
Molecular Weight: 211.28
Purity/Analysis Method: >98.0% (HPLC,N)
Form: Crystal
Melting point (°C): 248


Product Categories: Bioscience, Non-detergent Sulfobetaines (NDSB)
Other Product Names / Synonyms: 3-[(2-Hydroxyethyl)dimethylammonio]propane-1-sulfonate, NDSB 211

Formula: C₇H₁₇NO₄S
MW: 211.28 g/mol
Melting Pt: 248 °C
MDL Number: MFCD01320397
CAS Number: 38880-58-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=98626fbd21891eb26962881621f6caf65b993192&vid=bb89e08c253e71cb5aa9fd842237da592d0cfa2b&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,N)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR