6-Hydroxypyrimidine-4-carboxylic acid ≥98.0% (by HPLC, titration analysis)
Supplier: TCI America
Synonyms:
6-Hydroxy-4-pyrimidinecarboxylic acid, 1,6-Dihydro-6-oxo-4-pyrimidinecarboxylic acid
H1392-1G
H1392-200MG
TCH1392-1GEA
170.32
CAD
TCH1392-1G
TCH1392-200MG
6-Hydroxypyrimidine-4-carboxylic acid ≥98.0% (by HPLC, titration analysis)
6-Hydroxypyrimidine-4-carboxylic acid
CAS Number: 6299-87-2
MDL Number: MFCD00023267
Molecular Formula: C5H4N2O3
Molecular Weight: 140.10
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal
Melting point (°C): 267
Other Product Names / Synonyms: 1,6-Dihydro-6-oxo-4-pyrimidinecarboxylic Acid
Formula:
C₅H₄N₂O₃ MW: 140.0969 g/mol Melting Pt: 267 °C |
MDL Number:
MFCD00023267 CAS Number: 6299-87-2 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...