6-Hydroxypyridazine-3-carboxylic acid monohydrate ≥98.0%

Supplier: TCI America

Synonyms: 6-Hydroxy-3-pyridazinecarboxylic Acid Monohydrate, 1,6-Dihydro-6-oxopyridazine-3-carboxylic acid monohydrate

H1387-1G H1387-5G
TCH1387-1GEA 52.14 CAD
TCH1387-1G TCH1387-5G
6-Hydroxypyridazine-3-carboxylic acid monohydrate ≥98.0%
6-Hydroxypyridazine-3-carboxylic acid monohydrate

CAS Number: 306934-80-5
MDL Number: MFCD09064936
Molecular Formula: C5H4N2O3
Molecular Weight: 140.10
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal
Melting point (°C): 257

Formula: C₅H₆N₂O₄
MW: 158.11 g/mol
Melting Pt: 257 °C
MDL Number: MFCD09064936
CAS Number: 306934-80-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3e6eefbeb098cdb078eb93a9133919a2d2b3ccda&vid=3e6eefbeb098cdb078eb93a9133919a2d2b3ccda&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR