4-Hydroxy-3-methoxy-ɑ-methylbenzyl alcohol ≥98.0%

Supplier: TCI America

Synonyms: 2-Hydroxy-5-(1-hydroxyethyl)anisole, 4-(1-Hydroxyethyl)-2-methoxyphenol

H1386-1G H1386-5G
TCH1386-1GEA 464.52 CAD
TCH1386-1G TCH1386-5G
4-Hydroxy-3-methoxy-ɑ-methylbenzyl alcohol ≥98.0%
4-Hydroxy-3-methoxy-alpha-methylbenzyl alcohol

CAS Number: 2480-86-6
MDL Number: MFCD00238638
Molecular Formula: C9H12O3
Molecular Weight: 168.19
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 102


Other Product Names / Synonyms: 2-Hydroxy-5-(1-hydroxyethyl)anisole, 4-(1-Hydroxyethyl)-2-methoxyphenol

Formula: C₉H₁₂O₃
Melting Pt: 102 °C
MDL Number: MFCD00238638
CAS Number: 2480-86-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9b9781d8132051858e2e01d16c15202beab65e0c&vid=9b9781d8132051858e2e01d16c15202beab65e0c&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR