2'-Hydroxy-5'-nitroacetophenone ≥98.0% (by GC, titration analysis)

Supplier: TCI America

Synonyms: 1-(2-Hydroxy-5-nitrophenyl)ethanone

H1378-25G H1378-5G
TCH1378-25GEA 943.89 CAD
TCH1378-25G TCH1378-5G
2'-Hydroxy-5'-nitroacetophenone ≥98.0% (by GC, titration analysis)
2'-Hydroxy-5'-nitroacetophenone

CAS Number: 1450-76-6
MDL Number: MFCD00463816
Molecular Formula: C8H7NO4
Molecular Weight: 181.15
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Melting point (°C): 102

Formula: C₈H₇NO₄
MW: 181.1500 g/mol
Melting Pt: 102 °C
MDL Number: MFCD00463816
CAS Number: 1450-76-6

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR