(1-Methyl-1H-imidazol-2-yl)methanol ≥98.0% (by GC, titration analysis)

Supplier: TCI America

Synonyms: 1-Methyl-2-imidazolemethanol

H1370-1G H1370-5G
TCH1370-1GEA 102.54 CAD
TCH1370-1G TCH1370-5G
(1-Methyl-1H-imidazol-2-yl)methanol ≥98.0% (by GC, titration analysis)
(1-Methyl-1H-imidazol-2-yl)methanol

CAS Number: 17334-08-6
MDL Number: MFCD00964673
Molecular Formula: C5H8N2O
Molecular Weight: 112.13
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Melting point (°C): 117


Other Product Names / Synonyms: 1-Methylimidazol-2-methanol

Formula: C₅H₈N₂O
MW: 112.13 g/mol
Melting Pt: 117 °C
MDL Number: MFCD00964673
CAS Number: 17334-08-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=057704f2f60742379e213a9dfabc6744ce960358&vid=057704f2f60742379e213a9dfabc6744ce960358&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR