1,6-Hexanediol dimethacrylate ≥98.0% stabilised
Supplier: TCI America
Synonyms:
1,6-Bis(methacryloyloxy)hexane, Hexane-1,6-diyl bis(2-methylacrylate)
H1369-25G
H1369-500G
TCH1369-25GEA
74.42
CAD
TCH1369-25G
TCH1369-500G
1,6-Hexanediol dimethacrylate ≥98.0% stabilised
1,6-Hexanediol dimethacrylate
(stabilized with MEHQ)
CAS Number: 6606-59-3
MDL Number: MFCD00015425
Molecular Formula: C14H22O4
Molecular Weight: 254.33
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Flash Point (°C): 109
Specific Gravity (20/20): 1.00
Product Categories: Materials Chemistry, Monomers
Other Product Names / Synonyms: 1,6-Bis(methacryloyloxy)hexane
Formula:
C₁₄H₂₂O₄ Density: 1.00 (20 °C) Flash Pt: 109 °C |
MDL Number:
MFCD00015425 CAS Number: 6606-59-3 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...