4-(Methylsulfonyl)phenol ≥98.0% (by GC)

Supplier: TCI America

H1367-25G H1367-5G
TCH1367-25GEA 195.92 CAD
TCH1367-25G TCH1367-5G
4-(Methylsulfonyl)phenol ≥98.0% (by GC)
4-(Methylsulphonyl)phenol

CAS Number: 14763-60-1
MDL Number: MFCD00047806
Molecular Formula: C7H8O3S
Molecular Weight: 172.20
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 95


Product Categories: Synthetic Organic Chemistry, Sulfones
Other Product Names / Synonyms: 4-(Methylsulfonyl)phenol

Formula: C₇H₈O₃S
MW: 172.20 g/mol
Melting Pt: 95 °C
MDL Number: MFCD00047806
CAS Number: 14763-60-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8c06346cc11ce8638f32e9c9333170a0fd37dc18&vid=8c06346cc11ce8638f32e9c9333170a0fd37dc18&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR