2-Hydroxy-4'-(2-hydroxyethoxy)-2-methylpropiophenone ≥98.0% (by HPLC)

Supplier: TCI America

Synonyms: 2-Hydroxy-1-(4-(2-hydroxyethoxy)phenyl)-2-methylpropan-1-one

H1361-25G H1361-5G H1361-100G
TCH1361-25GEA 261.27 CAD
TCH1361-25G TCH1361-5G TCH1361-100G
2-Hydroxy-4'-(2-hydroxyethoxy)-2-methylpropiophenone ≥98.0% (by HPLC)
2-Hydroxy-4'-(2-hydroxyethoxy)-2-methylpropiophenone

CAS Number: 106797-53-9
MDL Number: MFCD00085267
Molecular Formula: C12H16O4
Molecular Weight: 224.26
Purity/Analysis Method: >98.0% (HPLC)
Form: Crystal
Melting point (°C): 91


Product Categories: Materials Chemistry, Polymerization Initiators
Other Product Names / Synonyms: 2-Hydroxy-1-[4-(2-hydroxyethoxy)phenyl]-2-methyl-1-propanone

Formula: C₁₂H₁₆O₄
MW: 224.2530 g/mol
Melting Pt: 91 °C
MDL Number: MFCD00085267
CAS Number: 106797-53-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=624863f2c7d9893da9c9e403334d89f84c34e415&vid=624863f2c7d9893da9c9e403334d89f84c34e415&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR