Harmol ≥98.0%

Supplier: TCI America

Synonyms: 7-Hydroxy-1-methyl-9H-pyrido[3,4-b]indole, 1-Methyl-9H-pyrido[3,4-b]indol-7-ol

H1360-1G H1360-200MG
TCH1360-1GEA 783.68 CAD
TCH1360-1G TCH1360-200MG
Harmol ≥98.0%
Harmol

CAS Number: 487-03-6
MDL Number: MFCD00834164
Molecular Formula: C12H10N2O
Molecular Weight: 198.23
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Melting point (°C): 323


Product Categories: Bioscience, Inhibitors (for Pharmacological Research), Crude Medicine Ingredients for Research and Experimental Use, Indole Alkaloids
Other Product Names / Synonyms: 7-Hydroxy-1-methyl-9H-pyrido[3,4-b]indole, 1-Methyl-9H-pyrido[3,4-b]indol-7-ol

Formula: C₁₂H₁₀N₂O
Melting Pt: 323 °C
MDL Number: MFCD00834164
CAS Number: 487-03-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=cfed60ac7dd12d6427ba5f316bd59009506792d2&vid=cfed60ac7dd12d6427ba5f316bd59009506792d2&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR