cis-4-Hydroxy-D(+)-proline ≥98.0%
Supplier: TCI America
Synonyms:
(R)-(+)-cis-4-Hydroxyproline, cis-4-Hydroxy-D-proline, H-D-cis-Hyp-OH, H-cis-D-Hyp-OH, D(+)-cis-4-Hydroxyproline
CAS Number: 2584-71-6
MDL Number: MFCD00005252
Molecular Formula: C5H9NO3
Molecular Weight: 131.13
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal
Melting point (°C): 248
Specific rotation [a]20/D: 58 deg (C=2, H2O)
Product Categories: Bioscience, Biologically-modified Amino Acids
Other Product Names / Synonyms: (2R,4R)-4-Hydroxypyrrolidine-2-carboxylic Acid, H-D-cis-Hyp-OH
Formula:
C₅H₉NO₃ MW: 131.13 Melting Pt: 248 °C |
MDL Number:
MFCD00005252 CAS Number: 2584-71-6 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...